missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 3,6-Di-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside 95.0+%, TCI America™

Catalog No. M16151G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M16151G Supplier TCI America Supplier No. M16151G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 129575-89-9
Molecular Formula C35H33NO8
Molecular Weight (g/mol) 595.648
InChI Key QKFDXGIKVGVBGT-PVEIOGNQSA-N
PubChem CID 11006483
IUPAC Name 2-[(2S,3R,4R,5S,6R)-5-hydroxy-2-(4-methoxyphenoxy)-4-phenylmethoxy-6-(phenylmethoxymethyl)oxan-3-yl]isoindole-1,3-dione
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)O)OCC4=CC=CC=C4)N5C(=O)C6=CC=CC=C6C5=O

Specifications

Color White
Quantity 1 g
Formula Weight 595.65
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 3,6-Di-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside
TSCA No
Recommended Storage Freezer