missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenyl 2,6-Di-O-benzyl-beta-D-galactopyranoside 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 113°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 466.53 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Methoxyphenyl 2,6-Di-O-benzyl-beta-D-galactopyranoside |
Sécurité et manipulation
| TSCA | No |
| Recommended Storage | Freezer |