missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenol 99.0+%, TCI America™
Supplier: TCI America M0123100G
Specifications
| 4-Methoxyphenol | |
| 57°C | |
| C7H8O2 | |
| 100 g | |
| NWVVVBRKAWDGAB-UHFFFAOYSA-N | |
| 4-methoxyphenol | |
| 9015 | |
| 124.14 | |
| Crystalline Powder |
| 150-76-5 | |
| 243°C | |
| MFCD00002332 | |
| mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum | |
| COC1=CC=C(O)C=C1 | |
| 124.14 | |
| CHEBI:69441 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 150-76-5 | |
| 124.14 | |
| NWVVVBRKAWDGAB-UHFFFAOYSA-N | |
| 9015 | |
| 4-methoxyphenol |
| C7H8O2 | |
| MFCD00002332 | |
| mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum | |
| CHEBI:69441 | |
| COC1=CC=C(O)C=C1 |
Safety and Handling
EINECSNumber : (3)-0567
RTECSNumber : SL7700000
TSCA : Yes