missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenol 99.0+%, TCI America™

Catalog No. p-7132005
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M012325G 25 g
M0123100G 100 g
M0123500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M012325G Supplier TCI America Supplier No. M012325G
Only null left

Chemical Identifiers

CAS 150-76-5
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002332
InChI Key NWVVVBRKAWDGAB-UHFFFAOYSA-N
Synonym mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum
PubChem CID 9015
ChEBI CHEBI:69441
IUPAC Name 4-methoxyphenol
SMILES COC1=CC=C(O)C=C1

Specifications

Melting Point 57°C
Boiling Point 243°C
Quantity 25 g
Formula Weight 124.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenol
EINECSNumber (3)-0567
RTECSNumber SL7700000
TSCA Yes