accessibility menu, dialog, popup

UserName

H3COOH

CAS RN 150-76-5

IUPAC Name: 4-methoxyphenol
Synonyms: methoxyphenol MEHQ P-guaiacol sodium salt 4-hydroxyanisole mechinolum P-hydroxyanisole mono methyl ether hydroquinone novo-dermoquinona db brand of 4-hydroxyanisole mequinolum
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: NWVVVBRKAWDGAB-UHFFFAOYSA-N
SMILES: COC1=CC=C(O)C=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (7)
Quantity 
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (3)
  • (2)
  • (3)
  • (3)
  • (8)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (3)
  • (7)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (5)
  • (5)
  • (3)