missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxyphenacyl Bromide 98.0+%, TCI America™

Catalog No. M10045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10045G 5 g
M100425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10045G Supplier TCI America Supplier No. M10045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2632-13-5
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00000201
InChI Key XQJAHBHCLXUGEP-UHFFFAOYSA-N
Synonym 2-bromo-4'-methoxyacetophenone, 2-bromo-1-4-methoxyphenyl ethanone, ethanone, 2-bromo-1-4-methoxyphenyl, p-methoxyphenacyl bromide, 4'-methoxyphenacyl bromide, 2-bromo-1-4-methoxyphenyl ethan-1-one, 4-methoxyphenacyl bromide, bromomethyl 4-methoxyphenyl ketone, alpha-bromo-4-methoxyacetophenone, 2-bromo-4-methoxyacetophenone
PubChem CID 4965
IUPAC Name 2-bromo-1-(4-methoxyphenyl)ethan-1-one
SMILES COC1=CC=C(C=C1)C(=O)CBr

Specifications

Melting Point 71°C
Quantity 5 g
UN Number 3261
Formula Weight 229.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Methoxyphenacyl Bromide