missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxydiphenylamine 98.0+%, TCI America™
$318.75 - $1275.02
Chemical Identifiers
| CAS | 1208-86-2 |
|---|---|
| Molecular Formula | C13H13NO |
| Molecular Weight (g/mol) | 199.25 |
| MDL Number | MFCD00228649 |
| InChI Key | OBHGSIGHEBGGFS-UHFFFAOYSA-N |
| PubChem CID | 14581 |
| IUPAC Name | 4-methoxy-N-phenylaniline |
| SMILES | COC1=CC=C(NC2=CC=CC=C2)C=C1 |
Chemical Identifiers
| 1208-86-2 | |
| 199.25 | |
| OBHGSIGHEBGGFS-UHFFFAOYSA-N | |
| 4-methoxy-N-phenylaniline |
| C13H13NO | |
| MFCD00228649 | |
| 14581 | |
| COC1=CC=C(NC2=CC=CC=C2)C=C1 |
Specifications
| 1208-86-2 | |
| White-Yellow | |
| C13H13NO | |
| 5 g | |
| COC1=CC=C(NC2=CC=CC=C2)C=C1 | |
| 199.25 | |
| 199.25 | |
| Crystalline Powder |
| 106°C | |
| 195°C | |
| MFCD00228649 | |
| OBHGSIGHEBGGFS-UHFFFAOYSA-N | |
| 4-methoxy-N-phenylaniline | |
| 14581 | |
| ≥98.0% (GC) | |
| 4-Methoxydiphenylamine |
Safety and Handling
EINECSNumber : (3)-3450
TSCA : No