missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxydiphenylamine 98.0+%, TCI America™
Supplier: TCI America M206925G
Specifications
| 4-Methoxydiphenylamine | |
| 106°C | |
| 195°C | |
| MFCD00228649 | |
| OBHGSIGHEBGGFS-UHFFFAOYSA-N | |
| 4-methoxy-N-phenylaniline | |
| 14581 | |
| ≥98.0% (GC) |
| 1208-86-2 | |
| White-Yellow | |
| C13H13NO | |
| 25 g | |
| COC1=CC=C(NC2=CC=CC=C2)C=C1 | |
| 199.25 | |
| 199.25 | |
| Crystalline Powder |
Chemical Identifiers
| 1208-86-2 | |
| 199.25 | |
| OBHGSIGHEBGGFS-UHFFFAOYSA-N | |
| 4-methoxy-N-phenylaniline |
| C13H13NO | |
| MFCD00228649 | |
| 14581 | |
| COC1=CC=C(NC2=CC=CC=C2)C=C1 |
Safety and Handling
EINECSNumber : (3)-3450
TSCA : No