accessibility menu, dialog, popup

UserName

CAS RN 1208-86-2

H3CONH

CAS RN 1208-86-2

IUPAC Name: 4-methoxy-N-phenylaniline
Synonyms: 4-methoxy-N-phenylbenzenamine
Molecular Weight (g/mol): 199.25
Molecular Formula: C13H13NO
InChi Key: OBHGSIGHEBGGFS-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results