missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxycarbonyl)cyclohexane-1-carboxylic acid, 97%

Catalog No. AAH5281406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5281406 5 g
AAH5281403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5281406 Supplier Thermo Scientific Chemicals Supplier No. H5281406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32529-79-6 | C9H14O4 | 186.207 g/mol

4-(Methoxycarbonyl)cyclohexane-1-carboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methoxycarbonyl)cyclohexane-1-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water (11 g/L at 25°C).

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 32529-79-6
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.207
MDL Number MFCD06797561
InChI Key ZQJNPHCQABYENK-UHFFFAOYSA-N
Synonym trans-1,4-cyclohexanedicarboxylic acid monomethyl ester, cis-4-methoxycarbonyl cyclohexanecarboxylic acid, trans-4-methoxycarbonyl cyclohexanecarboxylic acid, 4-methoxycarbonyl cyclohexanecarboxylic acid, 1r,4r-4-methoxycarbonyl cyclohexanecarboxylic acid, 4-methoxycarbonyl cyclohexane-1-carboxylic acid, 4-carbomethoxy-cyclohexane-1-carboxylic acid, trans-4-carbomethoxycyclohexane-1-carboxylic acid, 1r,4r-4-methoxycarbonyl cyclohexane-1-carboxylic acid, trans-1,4-cyclohexane dicarboxylic acid monomethyl ester
PubChem CID 296850
IUPAC Name 4-methoxycarbonylcyclohexane-1-carboxylic acid
SMILES COC(=O)C1CCC(CC1)C(=O)O

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water (11g/L at 25°C).
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 4-(Methoxycarbonyl)cyclohexane-1-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only