missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxybenzyl Alcohol 98.0+%, TCI America™
Supplier: TCI America M0107100G
Specifications
| 4-Methoxybenzyl Alcohol | |
| White | |
| C8H10O2 | |
| 100 g | |
| MSHFRERJPWKJFX-UHFFFAOYSA-N | |
| (4-methoxyphenyl)methanol | |
| 7738 | |
| 138.17 | |
| Crystalline Lumps |
| 105-13-5 | |
| 259°C | |
| MFCD00004653 | |
| 4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol | |
| COC1=CC=C(CO)C=C1 | |
| 138.17 | |
| CHEBI:86918 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 105-13-5 | |
| 138.17 | |
| MSHFRERJPWKJFX-UHFFFAOYSA-N | |
| 7738 | |
| (4-methoxyphenyl)methanol |
| C8H10O2 | |
| MFCD00004653 | |
| 4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol | |
| CHEBI:86918 | |
| COC1=CC=C(CO)C=C1 |
Safety and Handling
EINECSNumber : (3)-1054
RTECSNumber : DO8925000
TSCA : Yes