accessibility menu, dialog, popup

UserName

CAS RN 105-13-5

IUPAC Name: (4-methoxyphenyl)methanol
Synonyms: anise alcohol P-methoxybenzyl alcohol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: MSHFRERJPWKJFX-UHFFFAOYSA-N
SMILES: COC1=CC=C(CO)C=C1
Boiling Point 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (5)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)