missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl Alcohol 98.0+%, TCI America™

Catalog No. p-7131991
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M010725G 25 g
M0107100G 100 g
M0107500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M010725G Supplier TCI America Supplier No. M010725G
Only null left

Chemical Identifiers

CAS 105-13-5
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004653
InChI Key MSHFRERJPWKJFX-UHFFFAOYSA-N
Synonym 4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol
PubChem CID 7738
ChEBI CHEBI:86918
IUPAC Name (4-methoxyphenyl)methanol
SMILES COC1=CC=C(CO)C=C1

Specifications

Color White
Boiling Point 259°C
Quantity 25 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Methoxybenzyl Alcohol
EINECSNumber (3)-1054
RTECSNumber DO8925000
TSCA Yes