missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzoyl Chloride 99.0+%, TCI America™

Catalog No. M0721500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0721500G 500 g
M072125G 25 g
M0721100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0721500G Supplier TCI America Supplier No. M0721500G

Chemical Identifiers

CAS 100-07-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00000687
InChI Key MXMOTZIXVICDSD-UHFFFAOYSA-N
Synonym p-anisoyl chloride, benzoyl chloride, 4-methoxy, anisoyl chloride, 4-anisoyl chloride, p-methoxybenzoyl chloride, 4-methoxybenzoylchloride, 4-methoxybenzoic acid chloride, benzoyl chloride, p-methoxy, p-methoxybenzoic acid chloride, 4-methoxy-benzoyl chloride
PubChem CID 7477
IUPAC Name 4-methoxybenzoyl chloride
SMILES COC1=CC=C(C=C1)C(Cl)=O

Specifications

Color White-Yellow
Quantity 500 g
UN Number 1729
Formula Weight 170.59
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Methoxybenzoyl Chloride
EINECSNumber (3)-1684
RTECSNumber CA0270000
TSCA Yes
Recommended Storage Refrigerator