missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzophenone 98.0+%, TCI America™

Catalog No. M035725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M035725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M035725G Supplier TCI America Supplier No. M035725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 611-94-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00008403
InChI Key SWFHGTMLYIBPPA-UHFFFAOYSA-N
Synonym 4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, p-methoxybenzophenone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, phenyl p-anisyl ketone, 4-benzoylanisole, 4-methoxyphenyl phenylmethanone, unii-i4xj07373m, 4-methoxyphenyl-phenylmethanone
PubChem CID 69146
IUPAC Name (4-methoxyphenyl)-phenylmethanone
SMILES COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 60°C
Color White
Boiling Point 193°C
Quantity 25 g
Formula Weight 212.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxybenzophenone
EINECSNumber (9)-2128
RTECSNumber PC4962500
TSCA No