missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-N-phenylbenzamide, 97%

Catalog No. p-7040072
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5969003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5969003 Supplier Thermo Scientific Chemicals Supplier No. H5969003

CAS: 7465-88-5 | C14H13NO2 | 227.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7465-88-5
Molecular Formula C14H13NO2
Molecular Weight (g/mol) 227.26
MDL Number MFCD00025810
InChI Key XOJAJRFBOKCXPI-UHFFFAOYSA-N
Synonym 4-methoxy-n-phenyl-benzamide, n-phenyl-4-methoxybenzamide, anisanilid, p-anisanilide, 4-methoxyphenyl-n-benzamide, benzamide, 4-methoxy-n-phenyl
PubChem CID 346037
IUPAC Name 4-methoxy-N-phenylbenzamide
SMILES COC1=CC=C(C=C1)C(=O)NC1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 227.26
Percent Purity 97%
Chemical Name or Material 4-Methoxy-N-phenylbenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only