missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-4'-nitrostilbene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 133°C |
| Quantity | 5 g |
| Formula Weight | 255.27 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Methoxy-4′-nitrostilbene |
Safety and Handling
| TSCA | No |