missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-pyrrolin-2-one, 99%

Catalog No. AAL1934406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1934406 5 g
AAL1934403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1934406 Supplier Thermo Scientific Chemicals Supplier No. L1934406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69778-83-2 | C5H7NO2 | 113.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69778-83-2
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.116
MDL Number MFCD00071564
InChI Key TXKQBYYDTLOLHA-UHFFFAOYSA-N
Synonym 4-methoxy-3-pyrrolin-2-one, 4-methoxy-1h-pyrrol-2 5h-one, 4-methoxy-1,5-dihydro-2h-pyrrol-2-one, 4-methoxy-1,5-dihydropyrrol-2-one, 2h-pyrrol-2-one, 1,5-dihydro-4-methoxy, 4-methoxy-2,5-dihydro-1h-pyrrol-2-one, 4-methoxy-1,5-dihydropyrro-2-one, 1,5-dihydro-4-methoxy-2h-pyrrol-2-on, 4-methoxy-3-pyrroline-2-one
PubChem CID 574769
IUPAC Name 3-methoxy-1,2-dihydropyrrol-5-one
SMILES COC1=CC(=O)NC1

Specifications

Melting Point 130°C to 134°C
Quantity 5 g
Beilstein 1524505
Formula Weight 113.12
Percent Purity 99%
Chemical Name or Material 4-Methoxy-3-pyrrolin-2-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only