missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-nitroaniline, 97%

Catalog No. p-157652
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125960250 25 g Glass bottle
AC125961000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC125960250 Supplier Thermo Scientific Chemicals Supplier No. 125960250

CAS: 96-96-8 | C7H8N2O3 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96-96-8
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD00007152
InChI Key QFMJFXFXQAFGBO-UHFFFAOYSA-N
Synonym 2-nitro-4-methoxyaniline, 4-amino-3-nitroanisole, fast bordeaux gp, 2-nitro-p-anisidine, azobase nas, devol bordeaux b, bordeaux gp base, bordeaux gp salt, fast bordeaux gp base, fast bordeaux gp salt
PubChem CID 66793
ChEBI CHEBI:48973
IUPAC Name 4-methoxy-2-nitroaniline
SMILES COC1=CC(=C(C=C1)N)[N+](=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 123.0°C to 126.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3OC6H3(NO2)NH2
Quantity 25 g
Beilstein 13, 521
Solubility Information Solubility in water: slightly soluble
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material 4-Methoxy-2-nitroaniline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Harmful to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-547-2
RTECSNumber BY4415000
TSCA TSCA

RUO – Research Use Only