missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-1-naphthol 98.0+%, TCI America™
Supplier: TCI America M085425G
Specifications
| 4-Methoxy-1-naphthol | |
| 130°C | |
| MFCD00003976 | |
| 4-methoxy-1-naphthol, 1-hydroxy-4-methoxynaphthalene, 1-naphthalenol, 4-methoxy, 4-methoxynaphthol, 4-methoxy-1-naphthalenol, walrycin a, 4-methoxynaphth-1-ol, ursol grounding blue bl, 1-naphthol, 4-methoxy, acmc-209px1 | |
| COC1=CC=C(C2=CC=CC=C21)O | |
| 174.199 | |
| 174.20 | |
| Crystalline Powder |
| 84-85-5 | |
| C11H10O2 | |
| 25 g | |
| BOTGCZBEERTTDQ-UHFFFAOYSA-N | |
| 4-methoxynaphthalen-1-ol | |
| 66542 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 84-85-5 | |
| 174.199 | |
| BOTGCZBEERTTDQ-UHFFFAOYSA-N | |
| 66542 | |
| COC1=CC=C(C2=CC=CC=C21)O |
| C11H10O2 | |
| MFCD00003976 | |
| 4-methoxy-1-naphthol, 1-hydroxy-4-methoxynaphthalene, 1-naphthalenol, 4-methoxy, 4-methoxynaphthol, 4-methoxy-1-naphthalenol, walrycin a, 4-methoxynaphth-1-ol, ursol grounding blue bl, 1-naphthol, 4-methoxy, acmc-209px1 | |
| 4-methoxynaphthalen-1-ol |
Safety and Handling
EINECSNumber : (4)-0359
TSCA : Yes