missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-1-naphthol 98.0+%, TCI America™

Catalog No. p-7132407
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08545G 5 g
M085425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08545G Supplier TCI America Supplier No. M08545G

Chemical Identifiers

CAS 84-85-5
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.199
MDL Number MFCD00003976
InChI Key BOTGCZBEERTTDQ-UHFFFAOYSA-N
Synonym 4-methoxy-1-naphthol, 1-hydroxy-4-methoxynaphthalene, 1-naphthalenol, 4-methoxy, 4-methoxynaphthol, 4-methoxy-1-naphthalenol, walrycin a, 4-methoxynaphth-1-ol, ursol grounding blue bl, 1-naphthol, 4-methoxy, acmc-209px1
PubChem CID 66542
IUPAC Name 4-methoxynaphthalen-1-ol
SMILES COC1=CC=C(C2=CC=CC=C21)O

Specifications

Melting Point 130°C
Quantity 5 g
Formula Weight 174.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-1-naphthol
EINECSNumber (4)-0359
TSCA Yes