missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptopyridine, 96%

Catalog No. AC164710010
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164710010 1 g Glass bottle
AC164710100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC164710010 Supplier Thermo Scientific Chemicals Supplier No. 164710010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4556-23-4 | C5H5NS | 111.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4556-23-4
Molecular Formula C5H5NS
Molecular Weight (g/mol) 111.16
MDL Number MFCD00006422
InChI Key FHTDDANQIMVWKZ-UHFFFAOYSA-N
Synonym 4-mercaptopyridine, pyridine-4-thiol, 4-pyridinethiol, 4-thiopyridine, 4-pyridinethione, pyridine, 4-mercapto, 4-thiopyridone, 4 1h-pyridinethione, pyridine-4 1h-thione, 5-21-07-00167 beilstein handbook reference
PubChem CID 2723889
SMILES S=C1C=CNC=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 182°C to 189°C
Color Brown or Green-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula (C5H4N)SH
Quantity 1 g
Beilstein 21, II, 35
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone,toluene,methanol and ethanol
IUPAC Name 1,4-dihydropyridine-4-thione
Formula Weight 111.16
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 4-Mercaptopyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
EINECSNumber 224-926-1

RUO – Research Use Only