missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenylhydrazine hydrochloride, 98%

Catalog No. AAA1241606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1241606 5 g
AAA1241603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1241606 Supplier Thermo Scientific Chemicals Supplier No. A1241606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 118427-29-5 | C9H14N2 | 150.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118427-29-5
Molecular Formula C9H14N2
Molecular Weight (g/mol) 150.23
MDL Number MFCD00013461
InChI Key ZYATZFJUOXJFPY-UHFFFAOYSA-N
Synonym 4-isopropylphenylhydrazine hydrochloride, 4-isopropylphenyl hydrazine hydrochloride, 1-4-isopropylphenyl hydrazine hydrochloride, 4-propan-2-yl phenyl hydrazine hydrochloride, hydrazine, 4-1-methylethyl phenyl-, monohydrochloride, 4-isopropylphenylhydrazinehydrochloride, pubchem7533, acmc-1cbwc, 4-propan-2-ylphenyl hydrazine hydrochloride, ksc183e0j
PubChem CID 2733248
IUPAC Name (4-propan-2-ylphenyl)hydrazine;hydrochloride
SMILES CC(C)C1=CC=C(NN)C=C1

Specifications

Melting Point 203°C (decomposition)
Quantity 5 g
Formula Weight 186.69
Percent Purity 98%
Chemical Name or Material 4-Isopropylphenylhydrazine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only