missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzenesulfonyl chloride, 96%

Catalog No. AAL1042814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1042814 25 g
AAL1042806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1042814 Supplier Thermo Scientific Chemicals Supplier No. L1042814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54997-90-9 | C9H11ClO2S | 218.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54997-90-9
Molecular Formula C9H11ClO2S
Molecular Weight (g/mol) 218.70
MDL Number MFCD00041510
InChI Key CETRNHJIXGITKR-UHFFFAOYSA-N
Synonym 4-isopropylbenzenesulfonyl chloride, 4-isopropylbenzene-1-sulfonyl chloride, 4-isopropylbenzenesulfonylchloride, 4-propan-2-yl benzene-1-sulfonyl chloride, 4-isopropyl benzenesulfonyl chloride, 4-isopropyl-benzenesulfonyl chloride, 4-propan-2-yl benzenesulfonyl chloride, 4-isopropylbenzensulfonylchloride, 4-isopropylphenylsulfonyl chloride, p-isopropylbenzenesulfonyl chloride
PubChem CID 2734882
IUPAC Name 4-propan-2-ylbenzenesulfonyl chloride
SMILES CC(C)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Density 1.22
Boiling Point 142°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.539
Quantity 25 g
UN Number UN3265
Beilstein 1103862
Sensitivity Moisture sensitive
Formula Weight 218.7
Percent Purity 96%
Chemical Name or Material 4-Isopropylbenzenesulfonyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only