missing translation for 'onlineSavingsMsg'
Learn More

4-Isocyanatobenzeneboronic acid pinacol ester, 95%

Catalog No. AAH53003MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53003MD 250 mg
AAH5300303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH53003MD Supplier Thermo Scientific Chemicals Supplier No. H53003MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 380430-64-8 | C13H16BNO3 | 245.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 380430-64-8
Molecular Formula C13H16BNO3
Molecular Weight (g/mol) 245.085
MDL Number MFCD03411942
InChI Key RHKRIDVZGAXYQE-UHFFFAOYSA-N
Synonym 2-4-isocyanatophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-isocyanatophenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-4-isocyanatophenyl-4,4,5,5-tetramethyl, 4-isocyanatophenylboronic acid, pinacol ester, 4-isocyanatophenyl boronic acid pinacol ester, 4-isocyanatobenzeneboronic acid pinacol ester, 4-isocyanatophenyl boronic acid, pinacol ester, 4-isocyanatophenyl boronic acid,pinacol ester, 2-4-isocyanato-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
PubChem CID 2773474
IUPAC Name 2-(4-isocyanatophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N=C=O

Specifications

Melting Point 162°C to 168°C
Quantity 250 mg
Formula Weight 245.08
Percent Purity 95%
Chemical Name or Material 4-Isocyanatobenzeneboronic acid pinacol ester
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only