missing translation for 'onlineSavingsMsg'
Learn More

4-Isocyanato-1-methyl-1H-indole, 97%, Thermo Scientific™

Catalog No. CC45806CB
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC45806CB 250mg
CC45806DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC45806CB Supplier Thermo Scientific Chemicals Supplier No. CC45806CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 887922-92-1
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD08690262
InChI Key TVGVHXVLMROUII-UHFFFAOYSA-N
Synonym 4-isocyanato-1-methyl-1h-indole, 1h-indole, 4-isocyanato-1-methyl, 1-methylindol-4-isocyanate, 1h-indole,4-isocyanato-1-methyl, 1-methyl-1h-indole-4-yl isocyanate
PubChem CID 18525783
IUPAC Name 4-isocyanato-1-methylindole
SMILES CN1C=CC2=C(C=CC=C21)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250mg
Formula Weight 172.19
Percent Purity 97%
Chemical Name or Material 4-Isocyanato-1-methyl-1H-indole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.