missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isobutylbenzoic Acid 99.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 144°C |
| Color | White |
| Quantity | 10 g |
| Formula Weight | 178.23 |
| Percent Purity | ≥99.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Isobutylbenzoic Acid |
Sécurité et manipulation
| EINECSNumber | (3)-1338 |
| RTECSNumber | DH3073800 |
| TSCA | No |