missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-o-xylene, 98+%

Catalog No. AAA1323230
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1323230 250 g
AAA1323209 10 g
AAA1323218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1323230 Supplier Thermo Scientific Chemicals Supplier No. A1323230
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 31599-61-8 | C8H9I | 232.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31599-61-8
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00040989
InChI Key CSFRCLYFVINMBZ-UHFFFAOYSA-N
Synonym 4-iodo-o-xylene, 1,2-dimethyl-4-iodobenzene, benzene, 4-iodo-1,2-dimethyl, 3,4-dimethyliodobenzene, 1-iodo-3,4-dimethylbenzene, 4-iodo-ortho-xylene, 3,4-dimethyl-1-iodobenzene, 4-iodo-orthoxylene, 4-iodo-0-xylene, pubchem3105
PubChem CID 141646
IUPAC Name 4-iodo-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)I)C

Specifications

Density 1.633
Quantity 250 g
Refractive Index 1.602
Beilstein 1905729
Boiling Point 231°C to 233°C
Formula Weight 232.07
Percent Purity ≥98%
Chemical Name or Material 4-Iodo-o-xylene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only