missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2-nitrotoluene 98.0+%, TCI America™

Catalog No. I09935G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I09935G Supplier TCI America Supplier No. I09935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41252-97-5
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.034
MDL Number MFCD00051090
InChI Key QLMRDNPXYNJQMQ-UHFFFAOYSA-N
Synonym 4-iodo-2-nitrotoluene, benzene, 4-iodo-1-methyl-2-nitro, pubchem15938, 2-nitro-4-iodotoluene, acmc-1apfk, maybridge1_008109, ksc493s9d, 4-iodo-1-methyl-2-nitro-benzene
PubChem CID 170481
IUPAC Name 4-iodo-1-methyl-2-nitrobenzene
SMILES CC1=C(C=C(C=C1)I)[N+](=O)[O-]

Specifications

Melting Point 61°C
Quantity 5 g
Formula Weight 263.03
Percent Purity ≥98.0% (GC)
Physical Form Powder
Chemical Name or Material 4-Iodo-2-nitrotoluene