missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2-methoxyaniline, 97%

Catalog No. AC444800010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444800010 1 g
AC444800050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444800010 Supplier Thermo Scientific Chemicals Supplier No. 444800010

CAS: 338454-80-1 | C7H8INO | 249.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 338454-80-1
Molecular Formula C7H8INO
Molecular Weight (g/mol) 249.05
InChI Key AEPCMLLYVXZOLQ-UHFFFAOYSA-N
Synonym 4-iodo-2-methoxy-phenylamine, 2-amino-5-iodoanisole, benzenamine, 4-iodo-2-methoxy, 4-iodo-o-anisidine, 2-methoxy-4-iodoaniline, acmc-1aizp, 2-amino-5-iodoanisole 4-iodo-2-methoxyaniline
PubChem CID 46737999
IUPAC Name 4-iodo-2-methoxyaniline
SMILES COC1=C(C=CC(=C1)I)N

Specifications

Melting Point 48°C to 50°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 249.05
Percent Purity 97%
Chemical Name or Material 4-Iodo-2-methoxyaniline
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only