missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphthalonitrile 97.0+%, TCI America™

Numéro de catalogue. H08131G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H08131G 1 g
H08135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H08131G Fournisseur TCI America Code fournisseur H08131G
Il en reste null

Identifiants chimiques

CAS 30757-50-7
Molecular Formula C8H4N2O
Molecular Weight (g/mol) 144.13
MDL Number MFCD00191653
InChI Key FTVOPKROFUTOKY-UHFFFAOYSA-N
Synonym 3,4-Dicyanophenol
PubChem CID 3875765
IUPAC Name 4-hydroxybenzene-1,2-dicarbonitrile
SMILES OC1=CC(C#N)=C(C=C1)C#N

Spécifications

Melting Point 212°C
Quantity 1 g
Formula Weight 144.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxyphthalonitrile