missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphthalic Acid 98.0+%, TCI America™

Catalog No. H06095G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H06095G Supplier TCI America Supplier No. H06095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 610-35-5
Molecular Formula C8H6O5
Molecular Weight (g/mol) 182.131
MDL Number MFCD00013984
InChI Key MWRVRCAFWBBXTL-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, 4-hydroxy, 4-hydroxybenzene-1,2-dicarboxylic acid, 4-hydroxy-1,2-benzenedicarboxylic acid, 3,4-dicarboxyphenol, 4-hydroxy-phthalic acid, acmc-1avol, ksc357m7h, 4-hydroxyphthalic acid, 4-hydroxy-1,2-benzenedicarboxylicacid, 1,2-benzenedicarboxylicacid, 4-hydroxy
PubChem CID 11881
ChEBI CHEBI:27600
IUPAC Name 4-hydroxyphthalic acid
SMILES C1=CC(=C(C=C1O)C(=O)O)C(=O)O

Specifications

Melting Point 198°C
Quantity 5 g
Formula Weight 182.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxyphthalic Acid