missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphthalic acid, 98%, Thermo Scientific™

Catalog No. AC412130250
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC412130250 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC412130250 Supplier Thermo Scientific Chemicals Supplier No. 412130250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 610-35-5
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H6O5
Quantity 25g
Synonym 1,2-benzenedicarboxylic acid, 4-hydroxy, 4-hydroxybenzene-1,2-dicarboxylic acid, 4-hydroxy-1,2-benzenedicarboxylic acid, 3,4-dicarboxyphenol, 4-hydroxy-phthalic acid, acmc-1avol, ksc357m7h, 4-hydroxyphthalic acid, 4-hydroxy-1,2-benzenedicarboxylicacid, 1,2-benzenedicarboxylicacid, 4-hydroxy
Solubility Information Solubility in water: insoluble.
InChI Key MWRVRCAFWBBXTL-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1O)C(=O)O)C(=O)O
IUPAC Name 4-hydroxyphthalic acid
Molecular Weight (g/mol) 182.13
PubChem CID 11881
ChEBI CHEBI:27600
Formula Weight 182.13
Percent Purity 98%
Chemical Name or Material 4-Hydroxyphthalic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 210-221-6

RUO – Research Use Only