missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylarsonic Acid 98.0+%, TCI America™

Catalog No. H020125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H020125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H020125G Supplier TCI America Supplier No. H020125G
Only null left

Chemical Identifiers

CAS 98-14-6
Molecular Formula C6H7AsO4
Molecular Weight (g/mol) 218.04
MDL Number MFCD00045707
InChI Key RSRFURJMCNJVOW-UHFFFAOYSA-N
Synonym 4-Hydroxybenzenearsonic Acid
PubChem CID 7373
IUPAC Name (4-hydroxyphenyl)arsonic acid
SMILES C1=CC(=CC=C1O)[As](=O)(O)O

Specifications

Melting Point 174°C
Color White
Quantity 25 g
UN Number 3465
Formula Weight 218.04
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxyphenylarsonic Acid
RTECSNumber CY5075000
TSCA No