Learn More
4-Hydroxyphenylacetic acid, 99%
CAS: 156-38-7 | C8H8O3 | 152.149 g/mol
Supplier: Thermo Scientific Chemicals A1501822
Description
It is used as a reagent in the acylation of phenols and amines. It is also used for fluorometric determination of oxidative enzymes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a reagent in the acylation of phenols and amines. It is also used for fluorometric determination of oxidative enzymes.
Solubility
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
| 4-Hydroxyphenylacetic acid | |
| 148°C to 150°C | |
| MFCD00004347 | |
| 1448766 | |
| Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water. | |
| C1=CC(=CC=C1CC(=O)O)O | |
| 152.149 | |
| CHEBI:18101 | |
| 99% |
| 156-38-7 | |
| C8H8O3 | |
| 100 g | |
| 4-hydroxyphenylacetic acid, 2-4-hydroxyphenyl acetic acid, p-hydroxyphenylacetic acid, 4-hydroxyphenyl acetic acid, 4-hydroxyphenylacetate, 4-hydroxybenzeneacetic acid, benzeneacetic acid, 4-hydroxy, p-hydroxyphenyl acetic acid, parahydroxy phenylacetic acid, 4-carboxymethylphenol | |
| XQXPVVBIMDBYFF-UHFFFAOYSA-N | |
| 2-(4-hydroxyphenyl)acetic acid | |
| 127 | |
| 152.15 |
Chemical Identifiers
| 156-38-7 | |
| 152.149 | |
| XQXPVVBIMDBYFF-UHFFFAOYSA-N | |
| 127 | |
| 2-(4-hydroxyphenyl)acetic acid |
| C8H8O3 | |
| MFCD00004347 | |
| 4-hydroxyphenylacetic acid, 2-4-hydroxyphenyl acetic acid, p-hydroxyphenylacetic acid, 4-hydroxyphenyl acetic acid, 4-hydroxyphenylacetate, 4-hydroxybenzeneacetic acid, benzeneacetic acid, 4-hydroxy, p-hydroxyphenyl acetic acid, parahydroxy phenylacetic acid, 4-carboxymethylphenol | |
| CHEBI:18101 | |
| C1=CC(=CC=C1CC(=O)O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-851-3
RTECSNumber : AI2680000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only