accessibility menu, dialog, popup

UserName

OHOOH

CAS RN 156-38-7

IUPAC Name: 2-(4-hydroxyphenyl)acetic acid
Synonyms: 4-hydroxyphenylacetic acid P-hydroxyphenylacetic acid parahydroxyphenylacetate (4-hydroxy-phenyl)-essigsaeure parahydroxy phenylacetic acid parahydroxy phenylacetate
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: XQXPVVBIMDBYFF-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(O)C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)