missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylacetic acid, 99%

Catalog No. p-7022307
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1501814 25 g
AAA1501822 100 g
AAA1501836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1501814 Supplier Thermo Scientific Chemicals Supplier No. A1501814

CAS: 156-38-7 | C8H8O3 | 152.149 g/mol

It is used as a reagent in the acylation of phenols and amines. It is also used for fluorometric determination of oxidative enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent in the acylation of phenols and amines. It is also used for fluorometric determination of oxidative enzymes.

Solubility
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 156-38-7
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00004347
InChI Key XQXPVVBIMDBYFF-UHFFFAOYSA-N
Synonym 4-hydroxyphenylacetic acid, 2-4-hydroxyphenyl acetic acid, p-hydroxyphenylacetic acid, 4-hydroxyphenyl acetic acid, 4-hydroxyphenylacetate, 4-hydroxybenzeneacetic acid, benzeneacetic acid, 4-hydroxy, p-hydroxyphenyl acetic acid, parahydroxy phenylacetic acid, 4-carboxymethylphenol
PubChem CID 127
ChEBI CHEBI:18101
IUPAC Name 2-(4-hydroxyphenyl)acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)O

Specifications

Melting Point 148°C to 150°C
Quantity 25 g
Beilstein 1448766
Solubility Information Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material 4-Hydroxyphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-851-3
RTECSNumber AI2680000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only