missing translation for 'onlineSavingsMsg'
Learn More

(4-Hydroxyphenyl)diphenylmethanol 98.0+%, TCI America™

Catalog No. H10591G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H10591G Supplier TCI America Supplier No. H10591G
Only null left

Chemical Identifiers

CAS 15658-11-4
Molecular Formula C19H16O2
Molecular Weight (g/mol) 276.335
InChI Key QEIRHBKKYVWAOM-UHFFFAOYSA-N
Synonym 4-Hydroxytrityl Alcohol
PubChem CID 263208
IUPAC Name 4-[hydroxy(diphenyl)methyl]phenol
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=C(C=C3)O)O

Specifications

Melting Point 162°C
Color Yellow
Quantity 1 g
Formula Weight 276.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (4-Hydroxyphenyl)diphenylmethanol
TSCA No
Recommended Storage Refrigerator