missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxyphenyl benzoate, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
Spécifications
| Nom ou substance chimique | 4-Hydroxyphenyl benzoate |
| Perte à la dessication | 0.5% max. |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 97.5% min. (HPLC) |
| Quantité | 25g |
| Poids de formule | 214.22 |
| Pourcentage de pureté | 98% |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and kee |
| EINECSNumber | 219-479-4 |
RUO – Research Use Only