missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenyl benzoate, 98%, Thermo Scientific™

Numéro de catalogue. AC455730050
Modifier l'affichage
Click to view available options
Quantité:
5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
AC455730050 5g
AC455730250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC455730050 Fournisseur Thermo Scientific Chemicals Code fournisseur 455730050

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 2444-19-1
Formule moléculaire C13H10O3
Poids moléculaire (g/mol) 214.22
Numéro MDL MFCD00053304
Clé InChI JFAXJRJMFOACBO-UHFFFAOYSA-N
Synonyme hydroquinone monobenzoate, p-hydroxyphenyl benzoate, unii-zcy7o225cp, 4-benzoyloxyphenol, 1,4-diphenol, 1-benzoate, benzoic acid 4-hydroxyphenyl ester, 4-hydroxyphenyl benzoate, zcy7o225cp, 4-hydroxyphenylbenzoate, 4-hydroxyphenylbenzoat
PubChem CID 75549
Nom de l’IUPAC (4-hydroxyphenyl) benzoate
SOURIRES OC1=CC=C(OC(=O)C2=CC=CC=C2)C=C1

Spécifications

Nom ou substance chimique 4-Hydroxyphenyl benzoate
Perte à la dessication 0.5% max.
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 97.5% min. (HPLC)
Quantité 5g
Poids de formule 214.22
Pourcentage de pureté 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 219-479-4

RUO – Research Use Only