missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)tetrahydropyran, 98%

Catalog No. AAH6456806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6456806 5 g
AAH6456803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6456806 Supplier Thermo Scientific Chemicals Supplier No. H6456806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 14774-37-9 | C6H12O2 | 116.16 g/mol

Used as a reagent in the identification and optimization of pteridinone Toll-like receptor 7 agonists, which can be used for the oral treatment of viral hepatitis. 4-(Hydroxymethyl)tetrahydropyran is also a starting material for [1-[(Tetrahydro-2H-pyran-4-yl)methyl]-1H-indol-3-yl](2,2,3,3-tetramethylcyclopropyl)methanone which is a drug that acts as CB2 cannabinoid receptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a reagent in the identification and optimization of pteridinone Toll-like receptor 7 agonists, which can be used for the oral treatment of viral hepatitis. 4-(Hydroxymethyl)tetrahydropyran is also a starting material for [1-[(Tetrahydro-2H-pyran-4-yl)methyl]-1H-indol-3-yl](2,2,3,3-tetramethylcyclopropyl)methanone which is a drug that acts as CB2 cannabinoid receptor agonist.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 14774-37-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00457804
InChI Key YSNVSVCWTBLLRW-UHFFFAOYSA-N
Synonym tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol
PubChem CID 2773573
SMILES OCC1CCOCC1

Specifications

Boiling Point 111°C to 115°C (15 mmHg)
Flash Point 61°C (142°F)
Refractive Index 1.46
Quantity 5 g
Beilstein 103069
Solubility Information Slightly soluble in water.
IUPAC Name (oxan-4-yl)methanol
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 4-(Hydroxymethyl)tetrahydropyran
Hazard Category H227-H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H227-H315
Causes serious eye damage.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only