missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)pyridine-2-carboxylic acid, 95%

Catalog No. p-7031742
missing translation for 'changeView'
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3292103 1 g
AAH3292106 5 g
AAH3292114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. AAH3292103 Supplier Thermo Scientific Chemicals missing translation for 'supplierNo' H3292103
Only null left

CAS: 923169-37-3 | C7H7NO3 | 153.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 923169-37-3
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD08447083
InChI Key QMLYYPGJXCVEHA-UHFFFAOYSA-N
Synonym 4-hydroxymethyl picolinic acid, 4-hydroxymethyl pyridine-2-carboxylic acid, 4-hydroxymethyl pyridine-2-carboxylicacid
PubChem CID 22645434
IUPAC Name 4-(hydroxymethyl)pyridine-2-carboxylic acid
SMILES C1=CN=C(C=C1CO)C(=O)O

Specifications

Melting Point 178°C to 185°C (decomposition)
Quantity 1 g
Formula Weight 153.15
Percent Purity 95%
Chemical Name or Material 4-(Hydroxymethyl)pyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only