missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxydiphenylamine 98.0+%, TCI America™

Catalog No. A0470500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0470500G 500 g
A047025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0470500G Supplier TCI America Supplier No. A0470500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122-37-2
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00020142
InChI Key JTTMYKSFKOOQLP-UHFFFAOYSA-N
Synonym 4-hydroxydiphenylamine, 4-phenylamino phenol, p-anilinophenol, phenol, 4-phenylamino, p-hydroxydiphenylamine, p-oxydiphenylamine, phenyl-p-aminophenol, n-phenyl-p-aminophenol, 4-phenylaminophenol, p-hydroxydifenylamin
PubChem CID 31208
IUPAC Name 4-anilinophenol
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)O

Specifications

Melting Point 73°C
Boiling Point 165°C
Quantity 500 g
Formula Weight 185.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxydiphenylamine
EINECSNumber (3)-0878
RTECSNumber SJ6950000
TSCA Yes