missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxydiphenylamine, 98%

Catalog No. AAL1894518
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1894518 50 g
AAL1894509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1894518 Supplier Thermo Scientific Chemicals Supplier No. L1894518

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 122-37-2 | C12H11NO | 185.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-37-2
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00020142
InChI Key JTTMYKSFKOOQLP-UHFFFAOYSA-N
Synonym 4-hydroxydiphenylamine, 4-phenylamino phenol, p-anilinophenol, phenol, 4-phenylamino, p-hydroxydiphenylamine, p-oxydiphenylamine, phenyl-p-aminophenol, n-phenyl-p-aminophenol, 4-phenylaminophenol, p-hydroxydifenylamin
PubChem CID 31208
IUPAC Name 4-anilinophenol
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)O

Specifications

Melting Point 66°C to 71°C
Boiling Point 215°C to 218°C (12 mmHg)
Flash Point 209°C (408°F)
Odor Odorless
Quantity 50 g
Beilstein 511942
Formula Weight 185.23
Percent Purity 98%
Chemical Name or Material 4-Hydroxydiphenylamine
Hazard Category Danger
Hazard Statement GHS H Statement
H311-H318-H315-H335
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 204-538-9
RTECSNumber SJ6950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only