missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxychalcone, 97%

Catalog No. AAA1530714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1530714 25 g
AAA1530706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1530714 Supplier Thermo Scientific Chemicals Supplier No. A1530714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20426-12-4 | C15H12O2 | 224.26 g/mol

4-Hydroxychalcone is used as anti-angiogenesis, antineoplastic. It is also used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxychalcone is used as anti-angiogenesis, antineoplastic. It is also used as organic intermediates.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 20426-12-4
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.26
MDL Number MFCD00016488
InChI Key PWWCDTYUYPOAIU-DHZHZOJOSA-N
Synonym 4-hydroxychalcone, 4-hydroxy chalcone, 2e-3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, unii-jo97q47vbu, jo97q47vbu, 3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, e-3-4-hydroxyphenyl-1-phenyl-prop-2-en-1-one, 3-4-hydroxyphenyl-1-phenyl-2-propen-1-one, e-3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, ccris 2228
PubChem CID 5282361
ChEBI CHEBI:34423
IUPAC Name (E)-3-(4-hydroxyphenyl)-1-phenylprop-2-en-1-one
SMILES OC1=CC=C(\C=C\C(=O)C2=CC=CC=C2)C=C1

Specifications

Melting Point 183°C to 185°C
Quantity 25 g
Beilstein 2049155
Solubility Information Insoluble in water.
Formula Weight 224.26
Percent Purity 97%
Chemical Name or Material 4-Hydroxychalcone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
RTECSNumber UD1675000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only