Learn More
4-Hydroxybenzophenone, 98%
CAS: 1137-42-4 | C13H10O2 | 198.22 g/mol
Supplier: Thermo Scientific Chemicals A150960B
Description
4-Hydroxybenzophenone is used in the field of organic synthesis. It is used as pharmaceutical intermediate of clomifene citrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hydroxybenzophenone is used in the field of organic synthesis. It is used as pharmaceutical intermediate of clomifene citrate.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
| 4-Hydroxybenzophenone | |
| 132°C to 135°C | |
| C13H10O2 | |
| 1000 g | |
| 4-hydroxybenzophenone, 4-benzoylphenol, 4-hydroxyphenyl phenyl methanone, p-benzoylphenol, p-hydroxybenzophenone, methanone, 4-hydroxyphenyl phenyl, 4-hydroxyphenyl phenylmethanone, benzophenone, 4-hydroxy, unii-04r2lws0ms, 4'-hydroxybenzophenone | |
| NPFYZDNDJHZQKY-UHFFFAOYSA-N | |
| (4-hydroxyphenyl)-phenylmethanone | |
| 14347 | |
| 198.22 |
| 1137-42-4 | |
| 260°C to 262°C (24 mmHg) | |
| MFCD00002355 | |
| 777776 | |
| Insoluble in water. | |
| OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1 | |
| 198.22 | |
| CHEBI:34421 | |
| 98% |
Chemical Identifiers
| 1137-42-4 | |
| 198.22 | |
| NPFYZDNDJHZQKY-UHFFFAOYSA-N | |
| 14347 | |
| (4-hydroxyphenyl)-phenylmethanone |
| C13H10O2 | |
| MFCD00002355 | |
| 4-hydroxybenzophenone, 4-benzoylphenol, 4-hydroxyphenyl phenyl methanone, p-benzoylphenol, p-hydroxybenzophenone, methanone, 4-hydroxyphenyl phenyl, 4-hydroxyphenyl phenylmethanone, benzophenone, 4-hydroxy, unii-04r2lws0ms, 4'-hydroxybenzophenone | |
| CHEBI:34421 | |
| OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-507-1
RTECSNumber : PC4959775
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only