missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxybenzophenone, 98%
Description
4-Hydroxybenzophenone is used in the field of organic synthesis. It is used as pharmaceutical intermediate of clomifene citrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxybenzophenone is used in the field of organic synthesis. It is used as pharmaceutical intermediate of clomifene citrate.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
4-Hydroxybenzophenone is used in the field of organic synthesis. It is used as pharmaceutical intermediate of clomifene citrate.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
| Melting Point | 132°C to 135°C |
| Boiling Point | 260°C to 262°C (24 mmHg) |
| Quantity | 50 g |
| Beilstein | 777776 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 198.22 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Hydroxybenzophenone |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 214-507-1 |
| RTECSNumber | PC4959775 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only