accessibility menu, dialog, popup

UserName

CAS RN 1137-42-4

IUPAC Name: 4-benzoylphenol
Synonyms: 4-hydroxybenzophenone ion(1-) 4-hydroxybenzophenone 4-hydroxybenzophenone, ion(1-)
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: NPFYZDNDJHZQKY-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)