missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzeneboronic acid, 97%

Catalog No. AAL1559406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1559406 5 g
AAL1559403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1559406 Supplier Thermo Scientific Chemicals Supplier No. L1559406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71597-85-8 | C6H7BO3 | 137.93 g/mol

4-Hydroxybenzeneboronic acid is used for Suzuki-Miyaura coupling and Stille coupling and potential for introducing different alkyl groups, palladium-catalyzed aminocarbonylation and cross-coupling reactions, bio-supported palladium nanoparticles as phosphine-free catalyst for Suzuki reaction in water and Cu2O-catalyzed aerobic oxidative cross-coupling of tetrazoles. It is also used in preparation of rod-like dendronized polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzeneboronic acid is used for Suzuki-Miyaura coupling and Stille coupling and potential for introducing different alkyl groups, palladium-catalyzed aminocarbonylation and cross-coupling reactions, bio-supported palladium nanoparticles as phosphine-free catalyst for Suzuki reaction in water and Cu2O-catalyzed aerobic oxidative cross-coupling of tetrazoles. It is also used in preparation of rod-like dendronized polymers.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 71597-85-8
Molecular Formula C6H7BO3
Molecular Weight (g/mol) 137.93
MDL Number MFCD01074628
InChI Key COIQUVGFTILYGA-UHFFFAOYSA-N
Synonym 4-hydroxyphenyl boronic acid, 4-hydroxybenzeneboronic acid, 4-boronophenol, 4-hydroxyphenyl boranediol, p-hydroxyphenylboronic acid, 4-hydroxyphenyl-boronic acid, boronic acid, 4-hydroxyphenyl, pubchem1729, acmc-209ojb
PubChem CID 2734360
IUPAC Name (4-hydroxyphenyl)boronic acid
SMILES OB(O)C1=CC=C(O)C=C1

Specifications

Melting Point ∽285°C (decomposition)
Quantity 5 g
Beilstein 2963491
Solubility Information Soluble in water
Formula Weight 137.93
Percent Purity 97%
Chemical Name or Material 4-Hydroxybenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only