missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4-methyl-2-pentanone, ≥98.5% (GC), MilliporeSigma™ Supelco™

Numéro de catalogue. 111018316
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111018316 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111018316 Fournisseur Merck Emd Millipore Code fournisseur 788915ML
Il en reste null
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Spécifications

CAS 123-42-2
Vapor Pressure <1 mmHg (20°C)
Vapor Density 4 (vs. air)
Density 0.931 g/mL (at 25°C)
Flash Point 58°C - DIN 51755 Part 1
Physical Form Neat
Linear Formula (CH3)2 C(OH)CH2 COCH3
MDL Number MFCD00004471
Boiling Point 166°C
Synonym Diacetone alcohol
Type Analytical Standard
Quantity 5 mL
Formula Weight 116.16
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Afficher plus de résultats Afficher moins de résultats